Environmental Express Assembled Filter Funnel, PP, 0.1 um, 250/pk
Representative image only.
Environmental Express Item # WW-35200-35
Prevent sample-to-sample contamination to ensure accurate alpha particle spectra
  • Polypropylene construction withstands exposure to acids and bases
  • Choose convenient preassembled filter units or separate funnels and filters to accommodate any protocol
PRODUCT OPTIONS
₹87,900.00INR / Pkg of 1
SPECS & DESCRIPTION
Specifications & Description
  • Membrane MaterialPolypropylene (PP)
  • Pore Size (µm)0.1
  • Membrane Diameter (mm)25
  • Housing MaterialPolypropylene (PP)
  • Sample Volume (mL)68
  • DescriptionAssembled Filter Funnel, PP, 0.1 um, 250/pk
Key Features
  • Polypropylene construction withstands exposure to acids and bases
  • Choose convenient preassembled filter units or separate funnels and filters to accommodate any protocol
More About this Item
Total funnel capacity is 68 mL and filter diameter is 25 mm. Funnel stem fits a standard rubber stopper hole. Easily remove the filter to analyze filtrate.
Assembled filter funnels with filters include a top cap, filter support and filter.