Environmental Express Air Sampling Quartz Fiber Filters, Heat Treated, 37 mm; 100/Pk
Representative image only.
Environmental Express Item # WW-35209-45
Ideal for air particle monitoring and environmental testing
  • Laser cutting ensures a clean, fiber-free filter edge
  • Excellent temperature and chemical resistance
  • 3 month warranty
PRODUCT OPTIONS
₹6,400.00INR / Pkg of 100
SPECS & DESCRIPTION
Specifications & Description
  • MaterialQuartz Microfiber
  • Diameter (mm)37
  • Diameter (cm)3.7
  • Membrane SupportNo
  • DescriptionAir Sampling Quartz Fiber Filters, Heat Treated, 37 mm; 100/Pk
  • Warranty3 month
3 Month Warranty
Key Features
  • Laser cutting ensures a clean, fiber-free filter edge
  • Excellent temperature and chemical resistance
  • 3 month warranty
More About this Item
Quality filters to meet your industrial hygiene applications with excellent air permeability with minimal pressure drop.