Environmental Express ABF5000 AutoBlock® Fill Filling Station
Representative image only.
Environmental Express Item # WW-35250-00
Automate the most dangerous step of sample prep!
  • Fit this compact filler in a fume hood for safe and clean operation
  • Intuitive and user-friendly operation through touchscreen or PC
  • Assemble this modular unit within an hour
  • Works with existing HotBlock®, HotBlock® Pro, and HotBlock® 200 digestion systems
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Capacity (mL)100
  • Length (\")22
  • Graduation Interval (mL)0.1
  • Width (InchShort)31
  • Height (in)27
  • Power (VAC)120 / 240
  • DescriptionAutoBlock® Fill Filling Station
  • Warranty1 year
1 Year WarrantyConformite EuropeenneMade in the U.S.A.
Key Features
  • Fit this compact filler in a fume hood for safe and clean operation
  • Intuitive and user-friendly operation through touchscreen or PC
  • Assemble this modular unit within an hour
  • Works with existing HotBlock®, HotBlock® Pro, and HotBlock® 200 digestion systems
  • 1 year warranty
More About this Item
Obtain consistent results and reduce preparation time with the AutoBlock Fill. This automated diluter/filler works with any HotBlock digestion system—well formats are already loaded into the software. Use the touchscreen, or connect to a PC, to quickly select which wells need to be filled.
Avoid caustic fumes—compact footprint easily fits into a fume hood or benchtop air filtration unit. Eliminate the most dangerous step in metals digestion—acid and reagent addition!