Energizer ECR2025BP Button cell batteries, 3 V, 6/pack (Eveready # ECR2025)

Representative image only.
Energizer Mfr # ECR2025BP – Item # WW-05944-05
Button cell battery for Digi-Sense™ Traceable® General-Purpose Timers
  • 1.5V
  • 6 pack
  • 145 mA hours

PRODUCT OPTIONS

Call For Price

SPECS & DESCRIPTION

Specifications & Description

  • Cell TypeLithium
  • Capacity155 mA hours
  • Voltage (V)3
  • Diameter (mm)20
  • Thickness (mm)2.5
  • DescriptionButton cell batteries, 3 V, 6/pack (Eveready # ECR2025)

Key Features

  • 1.5V
  • 6 pack
  • 145 mA hours

More About this Item

Since 1896 Energizer® batteries have been providing dependable portable power.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat