EMD Millipore UFC203024 Amicon Ultra Centrifugal Filters, 2.0 mL, 30K MWCO; pack of 24

Representative image only.
EMD Millipore Mfr # UFC203024 – Item # WW-29968-34
Low-binding regenerated cellulose membrane, fast processing, and >90% recoveries
  • Unique vertical housing
  • Designed with 2.0-mL volume and 30,000 Nominal molecular weight limit (NMWL)

PRODUCT OPTIONS

₹40,100.00INR / Pkg of 24

SPECS & DESCRIPTION

Specifications & Description

  • Tube Size (mL)2
  • Max Sample Volume (mL)2
  • Nominal Molecular Weight Limit (Mwl) (Dalton)30000
  • Max Retention Volume (µL)70
  • Min Retention Volume (µL)15
  • DescriptionAmicon Ultra Centrifugal Filters, 2.0 mL, 30K MWCO; pack of 24

Key Features

  • Unique vertical housing
  • Designed with 2.0-mL volume and 30,000 Nominal molecular weight limit (NMWL)

More About this Item

These units are premier tools for protein concentration and desalting. Typical applications include purifying macromolecular components from tissue culture extracts or cell lysates and concentrating biological samples.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat