EMD Millipore PLBC06210 Ultrafiltration Disc Membranes, 3k 10pk
Representative image only.
EMD Millipore Mfr # PLBC06210 – Item # WW-29949-14
Ideal for stirred cells and micropartition system
  • Tight microstructure ensures the highest possible retention with the lowest possible adsorption of protein, DNA, or other macromolecules
  • 1 year warranty
PRODUCT OPTIONS
Call For Price
SPECS & DESCRIPTION
Specifications & Description
  • Nominal Molecular Weight Limit (Mwl) (Dalton)3000
  • Diameter (mm)63.5
  • Diameter (cm)6.35
  • DescriptionUltrafiltration Disc Membranes, 3k 10pk
  • Warranty1 year
1 Year Warranty
Key Features
  • Tight microstructure ensures the highest possible retention with the lowest possible adsorption of protein, DNA, or other macromolecules
  • 1 year warranty
More About this Item
These high-recovery hydrophilic membranes are made of regenerated cellulose. Use when concentrating or desalting extremely dilute solutions or whenever your sample is hydrophobic.