EMD Millipore 5121 Amicon Stirred Cell Model 8010, 10 mL for 25 mm Filters; 1/Pk

Representative image only.
EMD Millipore Mfr # 5121 – Item # WW-29968-12
Five different sizes handle volumes from 3 to 400 mL
  • Feature high flow rates with solutions up to 10% macrosolute concentration
  • 1 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • Membrane Diameter (mm)25
  • Effective Membrane Area (cm²)4.1
  • Min Process Volume (mL)1
  • Max Process Volume (mL)10
  • Hold-Up Volume (mL)0.2
  • DescriptionAmicon Stirred Cell Model 8010, 10 mL for 25 mm Filters; 1/Pk
  • Warranty1 year
1 Year Warranty

Key Features

  • Feature high flow rates with solutions up to 10% macrosolute concentration
  • 1 year warranty

More About this Item

Use these stirred cells to concentrate, diafilter, and exchange buffers for macromolecule solutions including proteins, enzymes, antibodies, and viruses. Maximum operating pressure is 75 psi. Recommended operating pressure is 55 psi (< 10 psi when using a 100K MWCO membrane or greater). Autoclavable at 121°C for 30 minutes.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat