This item is now discontinued. We recommend the following as suggested replacements: WW-18100-23
Elma Lab Clean S10 Ultrasonic Cleaner Detergent, Mildly Acidic Foam-Inhibited; 25 L
Representative image only.
Elma Mfr # 800 0098 – Item # WW-59988-77
Biodegradable, concentrated cleaners with long shelf lives
  • Solutions specifically made for labware
SPECS & DESCRIPTION
Specifications & Description
  • FormatLiquid
  • Volume (gallons)6.6
  • Volume (Liters)25
  • ApplicationsManual or ultrasonic cleaning. Lab use.
  • DescriptionUltrasonic Cleaner Detergent, Mildly Acidic Foam-Inhibited; 25 L
Key Features
  • Solutions specifically made for labware
More About this Item
S10 is mildly acidic and foam inhibited—use to clean plasticware, glassware, and metal instruments. Great for removing lime scale (and inhibiting redepositing), aqueous emulsions, light mineral oil and greases, fingerprints, and dust.