Cole-Parmer AT4001 Fume Collection Tube for Micro-Kjeldahl Extraction Heaters
Representative image only.
Cole-Parmer Item # WW-03074-99
Ensure safety with fume collection
  • 1 year warranty
  • Previous part number AT4001
₹67,200.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • MaterialGlass
  • Compatible WithMicro-kjeldahl extraction heaters 03074-30, 03074-32, and 03074 -34
  • DescriptionFume Collection Tube for Micro-Kjeldahl Extraction Heaters
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
  • Previous part number AT4001
More About this Item
Use gas tube to safely collect and exhaust fumes when not operating the micro-Kjeldahl extraction heater under a fume hood.