Electrolab Electromagnetic Sieve Shaker. 230 VAC, 50 HZ
Representative image only.
Electrolab Mfr # EMS-8 – Item # WW-15940-29
Shaker combines a vertical motion with a rotation of the material for more uniform particle separations
  • 316 stainless steel sieves have no crevices to trap material
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionElectromagnetic Sieve Shaker. 230 VAC, 50 HZ
  • Warranty1 Year
1 Year Warranty
Key Features
  • 316 stainless steel sieves have no crevices to trap material
  • 1 year warranty
More About this Item
This electromagnetic sieve shaker is ideal for use as a particle profile/size analyzer in the pharmaceutical, food, and chemical industries. The combination of a vertical movement with a rotation of the material produces faster and more efficient sieving. User-friendly operation is supported by the 16 x 2 alphanumeric LCD. Select the operation mode, either continuous or intermittent. Program in the process time from 1 to 99 minutes and amplitude of vibration from 0.5 to 2.0 mm.
The top plate of the shaker has clearly marked calibration lines for visual inspection of the amplitude. Electromagnetic drive has no rotating parts making it maintenance free and extremely quiet in operation. Use the wet sieve attachment with suspensions that should not dry and with very fine powders that tend to clump up and clog sieve openings.
The compact unit doesn't take up valuable bench space. A special clamping device ensures a tight grip on the sieves, but allows for quick and easy removal and replacement of sieves. Shaker can hold up to eight 50-mm sieves. Anti-vibration mountings isolate vibrations from the work surface, further reducing noise levels.
Test Sieves include an Inspection and Compliance certificate and are laser-marked with serial number and specifications. The sieves are all 316 stainless steel with welded mesh and a jointless rim for easy stacking of sieves. Sieves are manufactured in accordance with ISO 3310:1-2000 standards. Sieves measure 50 mm H x 200 mm diameter.