Ecodyst EcoChyll S Tankless Evaporator; 115 VAC, 60 Hz

Representative image only.
Ecodyst Mfr # EC787 – Item # WW-42000-00
Replace conventional coolant-dependent condensers
  • Reach -20°C in under two minutes!
  • Compatible with any rotary evaporator
  • Tankless design—no antifreeze, dry ice, or water needed
  • Compact footprint saves precious bench space
  • -40 to -5°C temperature range with ±2° stability
  • 1 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • Display TypeDual-line digital
  • Condenser TypeRefrigerated
  • Joint Size35/20
  • Capacity3000 mL receiving (1000-mL flask included)
  • Tube Size1/2\" ID hose barb
  • Min Temperature (° C)-40
  • Max Temperature (° C)-5
  • Temperature Resolution0.1°C
  • Power (VAC)115
  • Power (watts)656
  • Power (Hz)60
  • Length (in)12
  • Width (in)10
  • Height (in)27
  • Length (cm)30.5
  • Width (cm)25.4
  • Height (cm)68.6
  • DescriptionTankless Evaporator; 115 VAC, 60 Hz
  • Warranty1 Year
1 Year Warranty

Key Features

  • Reach -20°C in under two minutes!
  • Compatible with any rotary evaporator
  • Tankless design—no antifreeze, dry ice, or water needed
  • Compact footprint saves precious bench space
  • -40 to -5°C temperature range with ±2° stability
  • 1 year warranty

More About this Item

The new Ecodyst EcoChyll S tankless evaporator is progressing rotary evaporation. The state-of-the-art refrigeration unit takes the place of standard water-, ice-, dry ice-, or antifreeze-dependent glass condensers. Reach stable sub-freezing temperatures in seconds!

EcoChyll’s patented refrigerated condenser sets a new standard for cooling samples and processes—all without the use of traditional coolants. No need to purchase antifreeze, dry ice, or worry about water taps. EcoChyll S eliminates the hassles and hazards associated with dry ice usage with continuously-available cooling. The EcoChyll is easy to use, saves time and money, and is worry-free—allowing you to to focus on more complex tasks.

Conventional rotary evaporator condensers are constructed of glass, which is a poor conductor of heat. However, this tankless evaporator has a robust, chemical-resistant metal condenser that can exceed -35 °C cooling temperatures. Rapidly reach low temperatures to condense vapors from samples/solvents to decrease evaporation time and increase productivity!

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat