Ebro 1340-5522 TFN 520-Smp Compact E/J/K/T Thermocouple Thermometer with Single Input
Representative image only.
Ebro Mfr # 1340-5522 – Item # WW-91210-39
Available in single- and dual- channel models
  • IP40-rated housing and keypad
  • 2 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Meter Measurement TypeThermocouple
  • Number Of Probe Channels1
  • Types Of Probes AcceptedType E, Type J, Type K, Type T
  • Min Temperature (° F)-330
  • Max Temperature (° F)2190
  • Min Temperature (° C)-200
  • Max Temperature (° C)1200
  • AccuracyType E: ±0.5°F between -58° to 374°F; Type J: ±0.5°F between -58° to 428°F; Type K: ±0.5°F between -140 to 482°F; Type T: ±0.5°F between -58° to 302°F [Type E: ±0.3°C between -50° to 190°C; Type J: ±0.3°C between -50° to 220°C; Type K: ±0.3°C between -100° to 250°C; Type T: ±0.3°C between -50° to 150°C]
  • ResolutionType E: 0.2°F between -140° to 302°F; Type J: 0.2°F between -140° to 374°F; Type K: 0.2°F between -330° to 482°F; Type T: 0.2°F between -58° to 428°F [Type E: 0.1°C between -100° to 150°C; Type J: 0.1°C between -100° to 190°C; Type K: 0.1°C between -200° to 250°C; Type T: 0.1°C between -50° to 220°C]
  • IP RatingIP40
  • Display Type3.5-digit LCD
  • Height (in)5
  • Depth (in)3/4
  • Battery Life5 years
  • Battery3v Battery (1)
  • DescriptionTFN 520-Smp Compact E/J/K/T Thermocouple Thermometer with Single Input
  • Warranty2 year
Two year warranty
Key Features
  • IP40-rated housing and keypad
  • 2 year warranty
More About this Item
These thermometers are constructed of ABS plastic and have an IP40 rating ideal for use in harsh environments. The use of an advanced internal microprocessor has selectable updating from 0.5 to 15 seconds produces an accuracy of ±0.5°F. The meters are multifunctional featuring max/min, Hold, °F/°C selectable units, adjustable sampling rate, large LCD, and auto power-off after inactivity to save on battery power. Available in single- and dual-channel, both capable of accepting any type of J, K, T, and E thermocouple probes with miniconnector. Dual-channel model also features T1, T2, and delta (T1–T2). Probes are sold separately.