Ebro 1340-5516 TFE510 Compact Thermocouple Thermometer with Remote Probe
Representative image only.
Ebro Mfr # 1340-5516 – Item # WW-91210-37
Ideal for use in harsh environments including the food industry
  • IP67-rated housing and keypad
  • Compact size allows for convenient storage
  • 2 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Meter Measurement TypeThermocouple
  • Number Of Probe Channels1
  • Types Of Probes AcceptedType T
  • Min Temperature (° F)-60
  • Max Temperature (° F)570
  • Min Temperature (° C)-50
  • Max Temperature (° C)300
  • Resolution0.2°F (0.1°C)
  • IP RatingIP67
  • Display Type3.5-digit LCD
  • Height (in)5
  • Depth (in)3/4
  • No. of Probe(s) Included1
  • Battery Life5 years
  • Battery3v Battery (1)
  • DescriptionTFE510 Compact Thermocouple Thermometer with Remote Probe
  • Warranty2 year
Two year warrantyIP 67
Key Features
  • IP67-rated housing and keypad
  • Compact size allows for convenient storage
  • 2 year warranty
More About this Item
This meter has a robust splash/impact-resistant ABS food-safe housing, with a large LCD which is dishwasher safe. The use of an advanced internal microprocessor with selectable updates from 0.5 to 15 seconds and a highly accurate type T thermocouple produces a high level of accuracy. The meter features the ability to adjust the sampling intervals, deg>F/°C selectable units, battery-life indicator, and auto shutoff.