Eagle M12Y Modular Quik-Assembly Safety Cabinet, 12 Gal., 1 Door, 2 Shelves - Yellow
Representative image only.
Eagle Mfr # M12Y – Item # WW-81773-22
Assembles in minutes with no tools required
  • Stores corrosive liquids safety
  • Easy to store
  • 1 year warranty
PRODUCT OPTIONS
₹59,400.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPE
  • Capacity (Gal)12
  • Number of Shelves2
  • Width (in)18
  • Height (in)36
  • Depth (InchShort)22
  • ColorYellow
  • Self-ClosingNo
  • Shipping Weight (lbs)45
  • DescriptionModular Quik-Assembly Safety Cabinet, 12 Gal., 1 Door, 2 Shelves - Yellow
  • Warranty1 Year
1 Year Warranty
Key Features
  • Stores corrosive liquids safety
  • Easy to store
  • 1 year warranty
More About this Item
Modular all-HDPE cabinets are designed to safely store corrosive or aggressive liquids in labs and industrial locations. Assembly is simple, and when not in use, cabinets can be broken down in minutes without tools and easily stored. The stand-alone cabinets can be assembled with the option of the lockable door opening to the right or left.