Eagle 945BIO Biohazardous Plastic Waste Can, Foot-Operated, 10 Gal
Representative image only.
Eagle Mfr # 945BIO – Item # WW-81772-22
Highly visible — alerts workers to potential biohazards
  • Resistant to most chemical compounds
  • Self-closing lid with foot lever
  • OSHA 29 CFR 1910.1030 compliant
  • FM approved
PRODUCT OPTIONS
₹22,100.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • MaterialHigh Density Polyethyene (HDPE)
  • Capacity (Gal)10
  • Capacity (Liters)37.85
  • Exterior Height (in)18
  • Exterior Height (cm)45.72
  • ApprovalFM
  • DescriptionRed Biohazard Waste Can, 10 gallon
Key Features
  • Resistant to most chemical compounds
  • Self-closing lid with foot lever
  • OSHA 29 CFR 1910.1030 compliant
  • FM approved
More About this Item
BioHazardous waste cans are constructed of red high-density polyethylene (HDPE) and offer safe temporary storage of potential biohazards. Warning sticker on the front of the can advises you of potential biohazards. Meets OSHA requirements for exposures to blood borne pathogens. Cans will not rust or dent, are resistant to most chemical compounds, have no seam for a leak proof construction, and feature self-closing lid with foot lever and flared base for added stability.
One hundred percent designed and manufactured in the USA.