Eagle 933FL Oily Waste Can, HDPE, Hands-Free, Self Closing, 6 Gal
Representative image only.
Eagle Mfr # 933FL – Item # WW-09417-22
Unique shape to ensure stability
  • Self-closing lid with foot lever
  • Resistant to most chemical compounds
  • leak proof construction
  • FM approved
₹16,500.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • MaterialHigh Density Polyethyene (HDPE)
  • Capacity (Gal)6
  • Capacity (Liters)22.71
  • Exterior Height (in)16
  • Exterior Height (cm)40.64
  • ApprovalFM
  • DescriptionCombustible Waste Container, HDPE, 6 Gallon
FM Approved
Key Features
  • Self-closing lid with foot lever
  • Resistant to most chemical compounds
  • leak proof construction
  • FM approved
More About this Item
Constructed of high density polyethylene (HDPE) the hands-free oily waste can is impact resistant, rustproof, non-corroding, and impervious to many liquid chemicals, solvents, and corrosives. It offers fire safe temporary storage of solvent rags, waste rags, shavings and other materials subject to spontaneous combustion. Features a self-closing lid with foot lever and a flared base for added stability.
Designed and manufactured in the USA.