Eagle 1423 Galvanized Steel Disposal Safety Can, Flame Arrester, 2.5 Gal
Representative image only.
Eagle Mfr # 1423 – Item # WW-81772-00
Provides safe temporary storage of liquid wastes
  • Wide opening for easy pouring
  • FM Approved
PRODUCT OPTIONS
₹34,900.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • MaterialSteel
  • Capacity (Gal)2.5
  • Capacity (Liters)9.46
  • Exterior Height (in)12
  • Exterior Diameter (in)11.25
  • Exterior Height (cm)30.48
  • Exterior Diameter (cm)28.58
  • ApprovalFM
  • DescriptionRed Steel Waste Disposal Safety Can, 2.5 gallon
FM Approved
Key Features
  • Wide opening for easy pouring
  • FM Approved
More About this Item
The galvanized steel disposal safety cans, featuring a red powder coat finish, provide an intermediate station for temporary storage of liquid wastes. The venting spout cap opens to a wide 80 degree angle and can be locked open for conveniently emptying various type containers. All cans are equipped with a frame arrestor. Cap assembly opens automatically to relieve excessive internal vapor pressures. The free-swinging handle maintains can balance.
Order metal disposal cans for flammable liquids.