This item is now discontinued. We recommend the following as suggested replacements: WW-06287-37
Dynalon Two-sided polyamide spatula, spoon/spatula
Representative image only.
Dynalon Mfr # 391925-0003 – Item # WW-06260-42
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Overall Length (in)7
  • Overall Length (cm)17.78
  • MaterialPolyamide
  • SterileNo
  • DescriptionTwo-sided polyamide spatula, spoon/spatula
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Strong polyamide spatulas are glass fiber reinforced. Good chemical resistance against solvents; poor resistance to acids or oxidizing agents. Choose either double spatula or spoon/spatula style. Flat spatula end measures 11⁄4" (3.2 cm) L x 3⁄4" (1.9 cm) W. Spoon end measures 11⁄4" (3.2 cm) L and holds 1⁄2 tsp (2.5 mL).