Dymo 1738345 Desktop Label Printer, Type 210D; Uses D1 Label Tapes
Representative image only.
Dymo Mfr # 1738345 – Item # WW-65533-00
Affordable, easy-to-use printers feature a large, graphical display
  • Horizontal and vertical printing
  • Numerous fonts, print styles, and patterns to choose from
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionDesktop Label Printer, Type 210D; Uses D1 Label Tapes
  • Warranty1 Year
1 Year WarrantyConformite EuropeenneCanadian Standards Association
Key Features
  • Horizontal and vertical printing
  • Numerous fonts, print styles, and patterns to choose from
  • 1 year warranty
More About this Item
Label printers feature economical thermal transfer printing — no ribbons, toner, or cartridges needed. Easily choose printing styles that are specifically used for labeling file folders, hanging files, CDs, equipment, and instruments. Printers feature an ergonomically designed QWERTY keyboard for error-free use.
LabelManager 210D features intuitive labeling and quick access keys for punctuation, currency symbols and diacriticals. Printer also features six font sizes, seven styles, eight box styles, and underline, as well as a nine-label memory and “save text” feature. Multiple label printing options save you time. Printer accepts three label widths: 1/4", 3/8" and 1/2”. Works with six AA batteries (not included) or optional AC adapter (not included). Order batteries and AC adapter below.
What''''''''s included: one 1/2” black-on-white D1 label cassette.
LabelManager 360D allows you to customize labels and add emphasis by choosing from three fonts, seven font sizes, seven text styles, and eight different box styles or underline. Printer also features a nine-label memory, a “save text” feature, and the ability to print up to 10 copies of the same label. Printer also allows quick access to special symbols including punctuation and currency. Printer uses Dymo D1 labels in 1/4", 3/8”, 1/2" and 3/4” widths, and is powered by a rechargeable lithium-ion battery, eliminating the need for batteries.
What’s included: one 1/2” black-on-white D1 label cassette, one lithium-ion battery, and charging cord.