DWK Life Sciences (Wheaton) W348923-B Mini Bottle Benchtop Roller Culture System, 100V Japan

Representative image only.
DWK Life Sciences (Wheaton) Mfr # W348923-B – Item # WW-29300-56
Use with standard incubators and cold rooms – small footprint is perfect for labs with limited space!
  • Self-lubricated bearings eliminate routine maintenance issues
  • Attach up to two additional deck kits for versatility depending on your applications
  • 1 year warranty
Often bought with…

SPECS & DESCRIPTION

Specifications & Description

  • Number of Bottles Accepted4
  • Min Bottle Diameter (mm)38
  • Max Bottle Diameter (mm)240
  • Max Bottle Length (mm)240
  • Bottle Speed (rpm)3 to 45 (38 mm bottle), 2 to 30 (60 mm bottle)
  • Width (in)12 1/4
  • Height (in)7 1/8
  • Length (in)12 3/4
  • Width (cm)31.1
  • Height (cm)18.1
  • Length (cm)32.4
  • Plug TypeJapan
  • Power (VAC)100
  • Power (watts)14
  • Power (Hz)50/60
  • DescriptionMini Bottle Benchtop Roller Culture System, 100V Japan
  • Warranty1 year
1 Year WarrantyConformite EuropeenneCanadian Standards AssociationUL Listed.

Key Features

  • Self-lubricated bearings eliminate routine maintenance issues
  • Attach up to two additional deck kits for versatility depending on your applications
  • 1 year warranty

More About this Item

Obtain better distribution of media and uniform gassing with these benchtop rollers, designed to continuously rotate cylindrical bottles at optimum speeds. Transistorized solid state circuitry allows exact speed control of the bottles and is designed to compensate against sudden or prolonged voltage changes. Roller shafts are mounted in self-lubricated nylon bearings, which eliminate many maintenance problems. Order bottles separately.

Mini bottle benchtop roller culture systems are designed for small-scale mixing and agitation using four 30 mL or larger serum bottles, 100-125 mL media bottles, or 38 x 200 mm culture tubes. These will accommodate bottles 38 to 60mm in diameter, up to 240mm long, and provides speeds of 3 to 45 rpm. Order up to two additional deck kits to increase your rolling capacity.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat