DWK Life Sciences (Wheaton) W216939 Amber Wide-Mouth Bottle, Polyvinyl-Lined PP Cap, 1 oz, 24/cs

Representative image only.
DWK Life Sciences (Wheaton) Mfr # W216939 – Item # WW-34632-41
  • Made of Type III Soda-Lime Glass
Often bought with…

SPECS & DESCRIPTION

Specifications & Description

  • Capacity (mL)30
  • Capacity (oz)1
  • MaterialSoda-Lime Glass
  • Cap MaterialPolyvinyl-lined Polypropylene
  • Cap / Stopper Size28-400
  • Cap ColorWhite
  • AutoclavableYes
  • Outside Diameter (OD) (cm)3.4
  • Overall Height (cm)7
  • DescriptionAmber Wide-Mouth Bottle, Polyvinyl-Lined PP Cap, 1 oz, 24/cs

Key Features

  • Made of Type III Soda-Lime Glass

More About this Item

Use these round glass bottles to store dry powders, soil samples and specimens. Ideal for general lab use or field sampling needs. Bottles include tamper evident labels that provide assurance that the bottle hasn't been tampered with in transit. Hazard communication labels are also included to communicate necessary information on the bottle. Caps are attached to the bottle; labels are not.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat