DWK Life Sciences (Wheaton) Glass Extraction Thimble for Micro Soxhlet Extraction Apparatus; 15 x 60 mm
Representative image only.
Advantec Mfr # W416788 – Item # WW-99036-50
Seamless, high-purity filters for extraction and air monitoring
  • 1 year warranty
PRODUCT OPTIONS
₹13,300.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • MaterialGlass
  • Height (mm)60
  • Diameter (mm)15
  • DescriptionExtraction thimble for soxhlet extraction glassware systems, 25 mL
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Thimble filters are used in soxhlet apparatus for dust sampling and for extracting soluble components using organic solvents. Thimbles are made of cellulose. Typical applications include analysis of fats, oils, grease, organics, pesticides, pollutants, and additives in plastic/rubber materials.
Cellulose thimbles are suitable for extraction of organic compounds and dust sampling. Lipid content is <0.1% by weight; thickness is 1.5 mm.
An extraction thimble is required to support the sample within the extractor—order extraction glassware separately.
Compatible for use with glassware system 99036-50.