DWK Life Sciences (Wheaton) 224372 Vial Decapper Pliers for 13 mm Seals
Representative image only.
DWK Life Sciences (Wheaton) Mfr # 224372 – Item # WW-98800-63
Easily remove seals
  • Made of steel with gray plastic-coated handles
  • 1 year warranty
₹18,700.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Cap Diameter (mm)11
  • DescriptionVial Decapper Pliers for 13 mm Seals
  • Warranty1 year
1 Year Warranty
Key Features
  • Made of steel with gray plastic-coated handles
  • 1 year warranty
More About this Item
Plier Decappers are an inexpensive and easy way to remove 13 mm seals. They are made of steel with gray plastic-coated handles. To remove a seal, simply secure the seal between the jaws, squeeze the handles and twist the seal.