DWK Life Sciences (Wheaton) 223684 Serum Vials, Clear Type I Glass, 3 mL; 144/Cs

Representative image only.
DWK Life Sciences (Wheaton) Mfr # 223684 – Item # WW-08916-01
Special design adds strength for freeze drying applications
  • Made from low extractable Type 1 borosilicate glass

SPECS & DESCRIPTION

Specifications & Description

  • Volume (mL)3
  • Tube MaterialBorosilicate Glass
  • Tube Bottom ShapeFlat
  • Tube ColorClear
  • Height (in)1 1/2
  • Outside Diameter (in)21/32
  • Height (mm)38
  • Outside Diameter (mm)17
  • DescriptionSerum Vials, Clear Type I Glass, 3 mL; 144/Cs

Key Features

  • Made from low extractable Type 1 borosilicate glass

More About this Item

The blowback feature in the neck and specially designed bottom radius for added strength
are important for lyophilization applications. Vials are packaged in shrink wrapped modules
to greatly reduce particulate contamination. Available in clear or amber glass. Clear vials
also conform to ASTM Type I and Class A requirements.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat