DWK Life Sciences (Kimble) Kimax Shellback burettes with dust cap 25 ml

Representative image only.
DWK Life Sciences (Kimble) Mfr # 17150F-25 – Item # WW-34542-50
Significantly increases your visibility and reading accuracy
  • Maintain fluid integrity within burette (buret) with dust cap
  • Designed from ASTM Specification E287 requirements
  • Designed with a 25 mL capacity
  • 1 year warranty

PRODUCT OPTIONS

₹30,700.00INR / Case of 1

₹13,500.00 - ₹20,200.00INR / Each

Time to recalibrate? Click on the calibration service to view details, then order.
Often bought with…

SPECS & DESCRIPTION

Specifications & Description

  • Capacity (mL)25
  • Graduation Interval (mL)0.1
  • Tolerance0.06
  • DescriptionShellback burettes with dust cap 25 ml
  • Warranty1 year
1 Year Warranty

Key Features

  • Maintain fluid integrity within burette (buret) with dust cap
  • Designed from ASTM Specification E287 requirements
  • Designed with a 25 mL capacity
  • 1 year warranty

More About this Item

Shellbach straight-bore burettes (burets) are designed with a blue line on a broader white band running vertically behind the scale to create an optical effect which makes the meniscus and line to appear to form a point. Read meniscus at the narrowest point of the blue line for best results. Scale is durable black ceramic enamel.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat