DWK Life Sciences (Kimble) Kimax Glass Tilting Dispenser, 50 mL; 1/ea
Representative image only.
DWK Life Sciences (Kimble) Mfr # 759300-0050 – Item # WW-07899-50
Features tilted head to allow accurate measurement
  • 1 year warranty
PRODUCT OPTIONS
₹85,200.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Capacity (mL)50
  • Joint Size Number29/42
  • DescriptionGlass Tilting Dispenser, 50 mL; 1/ea
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Manufactured entirely of glass; dispenser features a fixed volume dispensing head secured with a head. Flasks tilts back to fill to diseired volume, forward to dispense. Clamp is included.