DWK Life Sciences (Kimble) Kimax Allihn Condenser; 125 mL; 1/Pk
Representative image only.
DWK Life Sciences (Kimble) Mfr # 456000-0021 – Item # WW-99037-26
Choose the condenser by type and sample volume
  • Allihn condenser (aka bulb or reflux condenser) better suited to basic laboratory applications
  • 1 year warranty
PRODUCT OPTIONS
₹43,300.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Capacity (mL)125
  • Condenser TypeAllihn
  • Flask Joint Size34/45
  • Condenser Joint Size34/45
  • DescriptionAllihn Condenser; 125 mL; 1/Pk
  • Warranty1 year
1 Year Warranty
Key Features
  • Allihn condenser (aka bulb or reflux condenser) better suited to basic laboratory applications
  • 1 year warranty
More About this Item
Freidrichs condenser (aka spiral cold finger condenser) better suited to heavy or difficult applications.