DWK Life Sciences (Kimble) KimCote® Class A Volumetric Flask with Glass Stopper; 1000 mL

Representative image only.
DWK Life Sciences (Kimble) Mfr # KC28014-1000 – Item # WW-34524-37
KimCote™ plastic coating for added safety
  • Reduces the hazards of shattered glass and leakage of toxic or corrosive chemicals
  • Volumetric flasks are constructed of low extractable borosilicate glass
  • Volumetric flask features 1000 mL (32 oz) capacity with ±0.30 mL tolerance
  • Features 13.25" (33.8 cm) height
  • Includes no.22 glass stopper

PRODUCT OPTIONS

₹122,000.00INR / Case of 4

₹20,400.00 - ₹27,500.00INR / Each

for an additional
₹20,400.00INR / Each
for an additional
₹27,500.00INR / Each
Time to recalibrate? Click on the calibration service to view details, then order.

SPECS & DESCRIPTION

Specifications & Description

  • MaterialGlass
  • Capacity (mL)1000
  • Tolerance (mL)±0.3
  • Cap TypePennyhead stopper
  • Stopper Number Accepted22
  • Overall Height (cm)33.8
  • DescriptionVolumetric Flask with Glass Stopper; 1000 mL

Key Features

  • Reduces the hazards of shattered glass and leakage of toxic or corrosive chemicals
  • Volumetric flasks are constructed of low extractable borosilicate glass
  • Volumetric flask features 1000 mL (32 oz) capacity with ±0.30 mL tolerance
  • Features 13.25" (33.8 cm) height
  • Includes no.22 glass stopper

More About this Item

Volumetric plastic-coated flasks feature white ceramic enamel marking spots and glass stoppers. Calibrated "To Contain", flasks include a single graduation ring. Flasks are designed to meet ASTM Spec E288, Class A unserialized requirements. Autoclavable to 120°C (250°F).

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat