DWK Life Sciences (Kimble) Glass Paley Bottle, Cheese Cream Test, 9 gram Sample; 1/Pack
Representative image only.
DWK Life Sciences (Kimble) Mfr # 509-50 – Item # WW-34590-41
₹19,300.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • MaterialGlass
  • AutoclavableYes
  • Overall Height (in)6 1/2
  • DescriptionGlass Paley Bottle, Cheese Cream Test, 9 gram Sample; 1/Pack
More About this Item
Easily measure the percentage of butter fat content in your sample with these specialized bottles. The body of each bottle is a reaction chamber, while the graduated neck is a measuring device. Use the Babcock bottles for measuring liquids and small samples. The Paley bottles feature a large opening on the side of the body to easily accommodate larger or viscous samples. Replacement stoppers are available: please see accessories. All bottles measure 6 1/2" H.