DWK Life Sciences (Kimble) 73770-16150 Disposable Round Bottom Culture Tubes, 16 x 150, no markings, 1000/cs
Representative image only.
DWK Life Sciences (Kimble) Mfr # 73770-16150 – Item # WW-34580-56
Ideal for general bacteriological and tissue culture applications
  • Tubes are offered with or without white markings
PRODUCT OPTIONS
₹91,000.00INR / Case of 1000
SPECS & DESCRIPTION
Specifications & Description
  • Capacity (mL)20
  • MaterialGlass
  • Cap / Stopper Size15-415
  • Label TypeNone
  • AutoclavableYes
  • Outside Diameter (OD) (cm)1.6
  • Overall Height (cm)15
  • DescriptionDisposable Round Bottom Culture Tubes, 16 x 150, no markings, 1000/cs
Key Features
  • Tubes are offered with or without white markings
More About this Item
Manufactured of borosilicate glass, these disposable tubes are ideal for general bacteriological and tissue culture applications. Caps are sold separately.