DWK Life Sciences (Kimble) 653150-0125 Gas Sampling Bulb, glass, 125 mL, 1/ea
Representative image only.
DWK Life Sciences (Kimble) Mfr # 653150-0125 – Item # WW-10370-00
₹60,100.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPTFE
  • Volume (Liters)0.125
  • DescriptionGas Sampling Bulb, glass, 125 mL, 1/ea
More About this Item
Glass Gas Sampling Bulbs are designed for the collection and storage of environmental samples. Top cap with a PTFE® PTFE/silicone septum lets you withdraw analytical samples. Both ends have a 4-mm valve plug and are ground to accept 3⁄8" ferrules and hand compressible fittings for a gas-tight seal. GPI thread size: 13-425.