DuPont Tychem QC Hooded Coveralls, Elastic Wrist and Ankles Large Cs/12
Representative image only.
DuPont Mfr # QC127SYLLG001200 – Item # WW-86220-04
Reliable protection from minor chemical spills, splashes, or mists
SPECS & DESCRIPTION
Specifications & Description
  • SizeLarge
  • MaterialPolyethylene(PE)
  • ColorYellow
  • SeamSerged
  • ComplianceASTM F1460, F1671
  • DescriptionHooded Coveralls, Elastic Wrist and Ankles Large Cs/12
More About this Item
Tychem® QC combines the strength of Tyvek® fabric with a polyethylene quality coating—offering a tough yet flexible fabric that resists tears and punctures. Garment provides reliable protection for minor chemical spills where there is potential for mist or light splash. Lightweight fabric is also durable and resistant to tear and abrasion. Seams are serged or sewn, or bound for greater strength and barrier quality depending on garment choice. Bright yellow color provides contrast across a wide range of natural backgrounds for high visibility. Suits can be used in a variety of environments such as pharmaceutical manufacturing, food processing, chemical processing, petroleum refining, and pulp and paper manufacturing.