DuPont TY120SWH002500 Coveralls, Zipper Front, Serged Seams, X-Large; 1/Pk
Representative image only.
DuPont Mfr # TY120SWHXL002500 – Item # WW-86225-03
Ideal for asbestos removal, general clean-up and maintenance, paint spraying, laboratories, and dry particulate protection
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPolyethylene(PE)
  • SizeX-Large
  • ColorWhite
  • SeamSerged
  • ComplianceANSI/ISEA 101-1996
  • DescriptionCoveralls, Zipper Front, Serged Seams, X-Large; 1/Pk
More About this Item
Rugged Tyvek® fabric resists snags, tears, punctures, and abrasion. The nonwoven fabric provides protection against particulates. Tyvek is chemically and biologically inert, low linting, and static-free until laundered.
These white coveralls have a zippered front that makes them easy to put on and take off. Coveralls feature serged seams.
Compliance: Meets sizing requirements of ANSI/ISEA 101-1996
Features open wrists and ankles.