DuPont Pro/Clean Coveralls, medium, bulk packed
Representative image only.
DuPont Mfr # PC143SWHMD00250B – Item # WW-33684-26
SPECS & DESCRIPTION
Specifications & Description
  • SizeMedium
  • MaterialPolyethylene(PE)
  • ColorWhite
  • SeamSerged
  • ComplianceISO Class 6, 7, 8 (formely Class 1000, 10000, 100000)
  • DescriptionCoveralls, medium, bulk packed
More About this Item
Pro/Clean® Coveralls are made from a unique breathable composite material for critical environments. This fabric provides excellent particulate and light splash protection. Coveralls feature a zipper front, serged seams, and elastic wrists, ankles, and waist. White.
Compliance: ISO Class 6, 7, and 8 (formerly Class 1000, 10,000, and 100,000)