Dow Corning 704 Vacuum Pump Oil, Silicone-Based; 500 mL
Representative image only.
Dow Corning Mfr # 704BB – Item # WW-79754-10
₹44,500.00INR / Each
Cole-Parmer CP 20 Vacuum Pump Oil, 58 cSt, 1 gal
₹32,400.00INR / Gallon of 1
SPECS & DESCRIPTION
Specifications & Description
  • FormLiquid
  • Volume (mL)500
  • Packaging500 mL
  • DescriptionVacuum Pump Oil, Silicone-Based; 500 mL
More About this Item
Vacuum pump oil is formulated to ensure optimum pump performance and pump life.
Dow Corning® 704 is a single component oil for deep vacuum pumps (10-6 to 10-8 torr untrapped, and 10-10 to 10-11 torr trapped). Offers quick reaction to maximum vacuum levels, even after exposure to air within operating temperatures.