Disposable Hemacytometers 50/Box - Neubauer Improved Grid
Representative image only.
Mfr # DHC-N015 – Item # WW-79003-00
Precise fixed-depth counting chamber with superior accuracy and reproducibility
  • Single use
  • Light and unbreakable
  • No coverslips required
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionDisposable Hemacytometers 50/Box - Neubauer Improved Grid
  • Warranty1 year
1 Year Warranty
Key Features
  • Single use
  • Light and unbreakable
  • No coverslips required
  • 1 year warranty
More About this Item
Our disposable hemocytometer is made of durable plastics used for manual cell counting. It consists of surface-patterned two enclosed 100µm height chambers with two ports for sample injection. It is a significant improvement of the conventional glass hemocytometer. Single usage reduces the risk of exposure to potential infectious material, increases productivity by eliminating cleaning and work interruptions.
100 um height chambers.