Digi-Vac 201VLC Vacuum Gauge, 0.001 to 760 Torr; 1/8" NPT Process Connection
Representative image only.
Digi-Vac Mfr # 201VLC – Item # WW-07379-22
Designed for precise laboratory processes
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Accuracy±1 mTorr below 0.009 Torr, ±7% of reading from 0.010 to 0.099 Torr, ±10% of reading from 0.1 to 0.999 Torr, ±15% of reading from 1.0 to 9.9 Torr, ±50% of reading from 10 to 160 Torr, ±25% of reading above 160 Torr
  • Max Vacuum (\" Hg)30
  • Min Vacuum (\" Hg)0
  • Process Connection1/8\" NPT(M)
  • Unit Of MeasurementTorr
  • Display6-digit red LED, 0.38\" H
  • Battery Type100 to 240 VAC, 50/60 Hz
  • Switch outputNO or NC
  • OutputRS-232, SPDT
  • Media CompatibilityClean, dry, noncorrosive gases
  • Resolution0.001 Torr
  • DescriptionVacuum Gauge, 0.001 to 760 Torr; 1/8\" NPT Process Connection
  • Warranty1 year
1 Year WarrantyConformite EuropeenneNo Liquids
Key Features
  • 1 year warranty
More About this Item
This vacuum gauge is a rugged instrument that measures vacuum through 6 decades of pressure from 0.001 to 760 Torr, and includes the industry standard Varian 531 gauge tube sensor. The base unit can be used as a benchtop or panel mounted in a 1/8 DIN cutout and comes standard with a large bright LED display, RS232 output and 2 set point relays. The relays can be used to turn on and off devices such as valves at specific vacuum levels. In addition, the 215 allows you to select the units displayed: either Torr, mBar or kPa. When purchased with the optional VLC control, the Model 215 VLC controls a vacuum level at a particular set point between 30 milliTorr and 2000 milliTorr. The VLC option also has user adjustable PID variables fine tune vacuum level control. The vacuum gauge can also be ordered with an optional vacuum controller that controls levels between 30 mTorr and 2000 mTorr. Voltage output, plotting software and web connectivity are also available, contact an Application Specialist.