Digi-Sense Type J Industrial Direct Insert Ungrounded Thermocouple Probe 4" L
Representative image only.
Digi-Sense Item # WW-93800-04
Available in spring loaded and non-spring loaded styles
  • 1 year warranty
PRODUCT OPTIONS
₹13,500.00INR / Each
₹27,000.00 - ₹33,700.00INR / Each
for an additional
₹27,000.00INR / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Probe TypeJ
  • Min Temperature (° F)32
  • Max Temperature (° F)1330
  • Min Temperature (° C)0
  • Max Temperature (° C)732
  • JunctionUngrounded
  • Sheath material316 Stainless Steel
  • Handle MaterialAluminum
  • Sheath Length (in)4
  • Sheath Diameter (in)0.25
  • DescriptionType J Industrial Direct Insert Ungrounded Thermocouple Probe 4\" L
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Thermocouple Probes are ideal for use in industrial applications, chemical processes, production lines, ovens, furnaces, and vessels. The probes conductor is made of 18 gauge wire and is encased with Magnesium Oxide insulation, and has a 316 stainless steel sheath. Featuring a standard size aluminum protection head with a gasketed cap to seal out dust and moisture and houses a ceramic terminal block. Head has a 3/4" NPT(F) opening on the side for easy connection to electrical conduit.
Spring loaded probes: designed to bottom out when inserted in to a thermowell to produce an accurate reading. (Not designed for direct insertion in to medium)
Non-spring loaded probes: designed to be directly inserted into the medium being measured without the use of a thermowell.