This item is now discontinued. We recommend the following as suggested replacements: WW-18001-08
Digi-Sense Traceable Cryogenic Temperature Data Logger with Calibration
Representative image only.
Digi-Sense Item # WW-18005-06
Built-in display shows measurements and alarm duration – no need to download data first
  • Includes NIST-Traceable certificate from our A2LA accredited (1750.01) calibration laboratory
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Number of Channels1
  • Input TypeTemperature
  • Max Temperature (° F)161.6
  • Min Temperature (° F)-328
  • Max Temperature (° C)72
  • Min Temperature (° C)-200
  • Temperature Resolution (° F)0.1
  • Temperature Accuracy (° F)±0.4 (32 to 86), ±2 (all other ranges)
  • Temperature Resolution (° C)0.1
  • Temperature Accuracy (° C)±0.2 (0 to 30), ±1 (all other ranges)
  • Memory Size64,000 temperature readings with time and date stamp
  • Alarm0 to 99 for ALARM
  • Battery Type1 x 3V CR2450 battery (included)
  • Display TypeLCD
  • DescriptionTraceable Cryogenic Temperature Data Logger with Calibration
  • Warranty1 Year
1 Year WarrantyConformite EuropeenneIP 66 RatedNIST Traceable Calibration
Key Features
  • Includes NIST-Traceable certificate from our A2LA accredited (1750.01) calibration laboratory
  • 1 year warranty
More About this Item
These compact, lightweight data loggers are reusable and reprogrammable making them ideal for transportation monitoring. Unit comes preprogrammed with alarm setpoints at 2°C (low) and 8°C (high) and to set record temperature readings every 5 minutes and 33 seconds for 30 days of continuous monitoring. If the measurement falls outside of the user-determined alarm conditions for too long, "QUARANTINE" is shown on the display—perfect for monitoring environmental temperatures of refrigerated vaccines, chemicals, pharmaceuticals, and other perishable items.
The IP66 waterproof rating allows the units to be used in any location, even those that are damp or require washdown. Order the optional sleeves to provide other users with a quick reference of the logger settings including minimum, maximum, quarantine, logging interval and duration, start and stop date and time, and special instructions.
Software is required to download and analyze data or program individual units. One copy of the software may be used for multiple loggers and allows you to update settings including individual unit name/description, time zone settings, daylight savings time on/off, unit of measure (°F or °C), start delay, recording duration/sampling interval, quarantine/alarm settings, and user stop key enabling. Software and cradle sold separately.
Traceable® Products have an individually-numbered Traceable® Certificate provided with each unit, that assures accuracy from our ISO/IEC 17025:2017(1750.01) calibration laboratory accredited by A2LA. It indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMI) that are signatories to the CIPM Mutual Recognition Agreement. Each product goes through an intense vetting process before it can carry the Traceable name, thus saving you both time and money by not having to calibrate separately.
These Digi-Sense temperature trackers have an IP66 waterproof rating allowing them to be used in any location, even those that are damp or require washdown. Order the optional sleeves to provide other users with a quick reference of the logger settings including minimum, maximum, quarantine, logging interval and duration, start and stop date and time, and special instructions.
Software is required to download and analyze data, or program individual units. One copy of the software may be used for multiple loggers and allows you to update settings including individual unit name/description, time zone settings, daylight savings time on/off, unit of measure (°F or °C), start delay, recording duration/sampling interval, quarantine/alarm settings, and user stop key enabling.
Cryogenic Temperature Data Logger has a separate probe specifically designed for ultra low temperature measurements. The external, stainless steel temperature probe is 4.35" (110.5 mm) long and has a 0.14" (3.55 mm) diameter—perfect for monitoring cryo-materials, reagents, samples, pharmaceuticals, and frozen food items. If the sample is subject to temperatures outside the user-defined set points, "ALARM" is displayed on the large LCD along with the number of times set points were breached.
What's included: battery and individually serialized NIST-traceable calibration certificate.
Requires software and cable
Order the optional sleeves to provide other users with a quick reference of the logger settings including minimum, maximum, quarantine, logging interval and duration, start and stop date and time, and special instructions.