Digi-Sense Temperature Probe, Type J, 6" L, 1/8" Dia, Fiberglass, Standard Jack
Representative image only.
Digi-Sense Item # WW-90446-35
Davis Instruments can provide many additional configurations beyond those listed here
  • 316 stainless steel sheath
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Probe TypeJ
  • Connector TypeStandard ANSI
  • Sheath Length (in)6
  • Sheath Diameter (in)0.125
  • DescriptionTemperature Probe, Type J, 6\" L, 1/8\" Dia, Fiberglass, Standard Jack
  • Warranty1 year
1 Year Warranty
Key Features
  • 316 stainless steel sheath
  • 1 year warranty
More About this Item
Magnesium Oxide insulated thermocouples, commonly referred to as MgO thermocouples, are used in many process and laboratory applications. Many desirable characteristics make them an excellent choice for general and special purpose applications. They are available in all thermocouple element types, single or dual elements, a wide variety of sheath diameters / materials, rugged in nature, bendable, and have fairly high temperature ratings. The thermocouples listed are commonly used; however, Davis Inotek Instruments can provide many additional configurations beyond those listed.