Digi-Sense Temp-360 Single-Input Data Logging RTD Thermometer
Representative image only.
Digi-Sense Item # WW-91428-08
Heavy-duty thermometer offers versatility and accuracy in a compact design
  • 3-way hands-free operation available
  • Large easy-to-read screen with back light
  • Rugged armor with built-in stand—withstand the toughest conditions
  • Easy-to-use automatic field calibration ensures accurate readings
  • Probes are sold separately
  • 3 year warranty
PRODUCT OPTIONS
₹106,000.00INR / Each
₹27,000.00 - ₹33,700.00INR / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Meter Measurement TypeRTD
  • Number Of Probe Channels1
  • Types Of Probes AcceptedRTD
  • Min Temperature (° F)-330
  • Max Temperature (° F)2210
  • Min Temperature (° C)-201
  • Max Temperature (° C)1210
  • Accuracy±0.1°C (0.2°F) below -100°C/F, ±0.03°C (0.6°F) From -99.99 to 99.99°C/°F, ±0.1°C (0.2°F) From 100.0 to 199.9°C/°F, ±0.5°C (1.0°F) From 200.0 to 500.0°C/°F, ±1.0°C (1.9°F) From above 500.0 to 999.9°C/°F, ±3°C (6°F) From above 1000°C/°F
  • Resolution0.01°; 0.1° between -100.0° to -330° and 100.0° to 999.9°; 1° above 1000°
  • OutputUSB
  • Data LoggingYes
  • Memory Capacity2000 data points
  • IP RatingIP54
  • Low Battery IndicatorYes
  • Display Type1.5\" Display, 4-digit LCD
  • Height (in)7
  • Depth (in)4
  • No. of Probe(s) Included0
  • Battery Life400 hours
  • Battery3 AA (included)
  • DescriptionTemp360 Single-Input Data Logging RTD Thermometer
  • Warranty3 year
3 Year WarrantyConformite Europeenne
Key Features
  • 3-way hands-free operation available
  • Large easy-to-read screen with back light
  • Rugged armor with built-in stand—withstand the toughest conditions
  • Easy-to-use automatic field calibration ensures accurate readings
  • Probes are sold separately
  • 3 year warranty
More About this Item
This robust and reliable meter work in any environment from the lab to the production floor. The sturdy armored construction protects against knocks and jars. All models are ergonomically designed for both large and small hands alike. The sealed keypad and ABS case meet IP54 standards for splash resistance.
Meter requires a RTD probe with a Digi-Sense three-pin plug.
OAKTON’s exclusive “3-WAY HANDS-FREE” capability allows “hands-free” operation— use the built-in stand on a table top, attach the meter to metal objects through magnets inserted into the armor boot, or hang from a pipe or belt using the hook-and-loop strap. Meter comes standard with the integral stand; the “hands-free” option adds magnets and strap (order separately).
Choose the Digi-Sense® Temp-340 thermometer for real-time data logging with USB data output. This robust and reliable meter is ideal for use in food processing plants, research and science labs, building maintenance areas, and more! The sturdy armored construction protects against knocks and jars. Ergonomically designed for both large and small hands alike. The sealed keypad and ABS case meet IP54 standards for splash resistance. The meter has a large illuminated display, and a seven-button control panel that allows the user through easy-to-use menus to choose desired functionality.
Features include real-time data logging and a USB data output. The seven-button control panel and intuitive on-screen menu guides you in choosing the desired functionality. Additional meter features include Min/Max, Hold, and average functions; °F/°C selection; time stamp/clock; countdown timer; out-of-range alarm with audible/visible indicator; auto shutoff; and low-battery indicator. Order an optional AC adapter to conserve battery life during extended operations. Manually or automatically log up to 2000 sets of readings in real time. Logging interval is selectable from 1 second to 60 minutes.