Digi-Sense Temp-10 Single-Input Type K Thermocouple Thermometer
Representative image only.
Digi-Sense Item # WW-91428-01
Heavy-duty thermometer offers versatility and accuracy in a compact design
  • 3-way hands-free operation available
  • Large easy-to-read screen with back light
  • Rugged armor with built-in stand—withstand the toughest conditions
  • Easy-to-use automatic field calibration ensures accurate readings
  • Probes are sold separately
  • 3 year warranty
PRODUCT OPTIONS
₹65,500.00INR / Each
₹19,800.00 - ₹27,800.00INR / Each
for an additional
₹19,800.00INR / Each
for an additional
₹27,800.00INR / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Meter Measurement TypeThermocouple
  • Number Of Probe Channels1
  • Types Of Probes AcceptedType K
  • Min Temperature (° F)-418
  • Max Temperature (° F)2501
  • Min Temperature (° C)-250
  • Max Temperature (° C)1372
  • Accuracy±0.1% of reading +0.7°F; ±0.25% of reading +2.0°F above -238°F [±0.1% of reading +0.4°C; ±0.25% of reading +1.0°C above -150°C]
  • Resolution0.1°; 1° above 999.9° and below -199.9°
  • Low Battery IndicatorYes
  • Display Type1.6\" Display, 4-digit LCD
  • Height (in)7
  • Depth (in)4
  • No. of Probe(s) Included0
  • Battery Life700 hours
  • Battery3 AA (included)
  • DescriptionTemp10 Single-Input Type K Thermocouple Thermometer
  • Warranty3 year
3 Year WarrantyConformite Europeenne
Key Features
  • 3-way hands-free operation available
  • Large easy-to-read screen with back light
  • Rugged armor with built-in stand—withstand the toughest conditions
  • Easy-to-use automatic field calibration ensures accurate readings
  • Probes are sold separately
  • 3 year warranty
More About this Item
These robust and reliable meters work in any environment from the lab to the production floor. The sturdy armored construction protects against knocks and jars. All models are ergonomically designed for both large and small hands alike. The sealed keypad and ABS plastic case meet IP54 standards for splash resistance. Select from the three most popular thermocouple types to meet all your basic temperature measurement needs.
Digi-Sense's exclusive “3-WAY HANDS-FREE” capability allows “hands-free” use—use the built-in stand on a table top, attach the meter to metal objects through magnets inserted into the armor boot, or hang from a pipe or belt using the hook-and-loop strap. Meters come standard with the integral stand; the “hands-free” option adds magnets and strap (order separately).
Features include a large, easy-to-read illuminated display; an easy-to-use five button control panel; Min/Max and Hold functions; °F/°C selection; auto shutoff; and low-battery indicator. Each thermometer is individually factory calibrated. Automatic field calibration is easily performed: simply place the probe in a container packed with ice, fill with water, then press the “CAL” button—meter automatically recognizes the freezing point.