Digi-Sense Standard Receptacle Tester SKU 2025061
Representative image only.
Digi-Sense Item # WW-20250-61
The fast and effortless way to test the safety of your electrical socket outlets
  • Inspect common faulty wiring issues in main socket installations
  • Easily find faults with bright light indicators
  • 2 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • AC Min (Volts)110
  • AC Max (Volts)125
  • DisplayLED
  • Interface3 lights indicator
  • MeasurementCategory II
  • Safety RatingCAT II 125 V, pollution 2; UL STD.61010-1
  • DescriptionCircuit Tester
  • Warranty2 Years
Two year warrantyUL Listed.
Key Features
  • Inspect common faulty wiring issues in main socket installations
  • Easily find faults with bright light indicators
  • 2 year warranty
More About this Item
Check the safety of electrical socket outlets by detecting wiring faults and incorrect grounding. Plug the tester into a standard 3-wire outlet, and different light combinations will indicate common wiring faults found in electrical sockets. Select from two types: circuit tester or GFCI circuit breaker tester. For use in standard 110 to 125 VAC outlets.
For use on 110-125V AC receptacles only. This is not a comprehensive diagnostics device.
Caution: Incorrect use may result in damage to the device or its components.