Digi-Sense ST221SS-1 Surface Thermometer, 2.5" Face, 1-Spring Pipe Mount, 0 to 250 ° F
Representative image only.
Digi-Sense Item # WW-08107-25
Inexpensive way to easily measure surface temperature
  • For reference only, not intended for applications where a precise reading is required
  • Item is not recommended to be calibrated
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Min Temperature (° F)0
  • Max Temperature (° F)250
  • Division (° F)2
  • Dial Size (in)2.5
  • Dial / Case Material304 Stainless Steel
  • Stem Material304 Stainless Steel
  • DescriptionSurface Thermometer, 2.5\" Face, 1-Spring Pipe Mount, 0 to 250 ° F
  • Warranty1 year
1 Year Warranty
Key Features
  • For reference only, not intended for applications where a precise reading is required
  • Item is not recommended to be calibrated
  • 1 year warranty
More About this Item
Fast, reliable, method of measuring surface temperature of pipes and other rounded surfaces. The temperature sensor is protected from drafts and other heat sources by a barrier guaranteeing integrity of readings. Two different models are available which are designed to fit around a large range of diameter surfaces. The 2 ½” model has a rugged 304SS case and is supplied with a single 300 series SS spring for mounting rounded surfaces of 1” and smaller. Both models are supplied with a heat treated bi-metallic sensor.