Digi-Sense Single-Input Thermocouple Thermometer System with NIST-Traceable Calibration
Representative image only.
Digi-Sense Item # WW-20250-98
Perfect for the lab to production floor
  • Reliable measurements, guaranteed!
  • Choose from meter or meter system—both include calibration
  • Heavy duty construction to protect against knocks and jars.
  • Super large LCD for easy viewing of data
  • Min, Max, and Avg readings, for quick analysis of the object
  • Electronic Offset function allows compensation of probe errors
  • Bundled kit for convenience and savings
  • One (1) probe and NIST-Traceable calibration included!
  • Accepts a wide variety of other probes
  • 2 year warranty
PRODUCT OPTIONS
₹45,100.00INR / Pkg of 1
₹27,000.00 - ₹33,700.00INR / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Meter Measurement TypeThermocouple
  • Number Of Probe Channels1
  • Types Of Probes AcceptedJ,K,T,R,S
  • Min Temperature (° F)-328
  • Max Temperature (° F)2501
  • Min Temperature (° C)-200
  • Max Temperature (° C)1372
  • Accuracy±1% of reading + 1.8⁰F (1⁰C) below -148⁰F (-99.9⁰C); ±1% of reading 0.9⁰F (0.5⁰C) above -148⁰F (-99.9⁰C)
  • Resolution0.1⁰F (0.1⁰C) from 148 to 1832⁰F, (-99.9 to 999.9⁰C); 1⁰F (1⁰C) outside range
  • Data LoggingNo
  • Low Battery IndicatorYes
  • Display TypeLarge, dual, backlit LCD
  • Battery Life3000 hours
  • Battery1 9V (included)
  • DescriptionSingle-Input Thermocouple Thermometer System with NIST-Traceable Calibration
Two year warrantyConformite EuropeenneNIST Traceable Calibration
Includes
General-purpose Type K thermocouple probe, protective rubber boot with built-in stand, soft-sided carrying case with strap, one 9 V battery, and NIST-traceable calibration report with data supplied by InnoCal.
Key Features
  • Reliable measurements, guaranteed!
  • Choose from meter or meter system—both include calibration
  • Heavy duty construction to protect against knocks and jars.
  • Super large LCD for easy viewing of data
  • Min, Max, and Avg readings, for quick analysis of the object
  • Electronic Offset function allows compensation of probe errors
  • Bundled kit for convenience and savings
  • One (1) probe and NIST-Traceable calibration included!
  • Accepts a wide variety of other probes
  • 2 year warranty
More About this Item
The Digi-Sense Professional Thermocouple Thermometers offer fast response and laboratory accuracy. Features include automatic backlight, Max/Min readings, and automatic power-off. These thermometers provide precise measurements when accuracy is a must, delivering dependable results every time! Each model is packed with distinctive features for use in a wide array of applications. Robust and reliable constructed to protects against knocks and jars. These thermometers produce a high degree of accuracy, with a rapid response time, and exceptional battery life of up to 3000 hours.
Digi-Sense instrument are fully tested and calibrated to NIST-traceable standards, saving you hundreds of dollars and extra waiting time to calibrate separately.