Digi-Sense RTD Thermometer, 2-Input Data Logging, NIST-Traceable Calibration
Representative image only.
Digi-Sense Item # WW-20250-96
Perfect for the lab to production floor
  • Reliable measurements, guaranteed!
  • NIST-traceable certificate included
  • Heavy duty construction to protect against knocks and jars.
  • Super large LCD for easy viewing of data
  • Accepts a wide variety of probes
  • Min, Max, and Avg readings, for quick analysis of the object
  • Electronic Offset function allows compensation of probe errors
  • Data logging models record up to 16,000 measurements for expanded data analysis capability
  • Data logging models feature software that provides quick and easy data interpretation
  • Temperature differential plus Min, Max, and Avg readings, for quick analysis of the object
  • Relative time clock on Min, Max, and Avg provides a time reference for major events
  • USB Interface
  • 2 year warranty
PRODUCT OPTIONS
₹25,200.00INR / Each
Time to recalibrate?
Often Bought with…
SPECS & DESCRIPTION
Specifications & Description
  • Meter Measurement TypeRTD
  • Number Of Probe Channels2
  • Types Of Probes AcceptedRTD
  • Min Temperature (° F)-320
  • Max Temperature (° F)1562
  • Min Temperature (° C)-200
  • Max Temperature (° C)850
  • Accuracy±1.5⁰F (0.8⁰C) from -148 to 392⁰F (-99.9 to 199.9⁰C); ±2⁰F (1⁰C) ±0.5% of reading outside range
  • Resolution0.1⁰F (0.1⁰C) from 148 to 1562⁰F (-99.9 to 850⁰C); 1⁰F (1⁰C) outside range
  • Data LoggingYes
  • Memory Capacity16000 data points
  • AlarmYes
  • Low Battery IndicatorYes
  • Display TypeLarge, dual, backlit LCD
  • Battery Life300 hours
  • Battery1 9V (included)
  • DescriptionRTD Thermometer, 2-Input Data Logging, NIST Traceable Cal
Two year warrantyConformite EuropeenneNIST Traceable Calibration
Key Features
  • Reliable measurements, guaranteed!
  • NIST-traceable certificate included
  • Heavy duty construction to protect against knocks and jars.
  • Super large LCD for easy viewing of data
  • Accepts a wide variety of probes
  • Min, Max, and Avg readings, for quick analysis of the object
  • Electronic Offset function allows compensation of probe errors
  • Data logging models record up to 16,000 measurements for expanded data analysis capability
  • Data logging models feature software that provides quick and easy data interpretation
  • Temperature differential plus Min, Max, and Avg readings, for quick analysis of the object
  • Relative time clock on Min, Max, and Avg provides a time reference for major events
  • USB Interface
  • 2 year warranty
More About this Item
The Digi-Sense Professional Single-Input Thermometers offer fast response and laboratory accuracy. Features include automatic backlight, Max/Min readings, and automatic power-off. These thermometers provide precise measurements when accuracy is a must, delivering dependable results every time! Each model is packed with distinctive features for use in a wide array of applications. Robust and reliable constructed to protects against knocks and jars. These thermometers produce a high degree of accuracy, with a rapid response time, and exceptional battery life of up to 3000 hours.
Digi-Sense instrument are fully tested and calibrated to NIST-traceable standards, saving you hundreds of dollars and extra waiting time to calibrate separately.
Advanced features include data logging, USB interface, relative time clock, offset compensation,
temperature differential.