Cole-Parmer Reversible 7-Point Horizontal Temperature Label, 0C-32C/32-86F; 10/Pk
Representative image only.
Cole-Parmer Item # WW-09037-07
Each style covers a wide range of temperatures, for use in many different application
  • Non-toxic / non-hazardous temperature measuring alternative
  • Easy to read and unbreakable
  • 1 year warranty
PRODUCT OPTIONS
₹1,900.00INR / Pkg of 10
SPECS & DESCRIPTION
Specifications & Description
  • Number of Temperature Indicators7
  • Max temperature (° F)86
  • Max temperature (° C)30
  • Temperature Points (° F)32-86
  • Temperature Points (° C)0 - 30
  • Initial colorBlack
  • First color changeGreen (actual temperature); Brown (above actual); Blue (below actual)
  • Second color changeBlack
  • Height (in)1/2
  • Width (in)1 3/4
  • DescriptionReversible 7-Point Horizontal Temperature Label, 0C-32C/32-86F; 10/Pk
  • Warranty1 year
1 Year Warranty
Key Features
  • Non-toxic / non-hazardous temperature measuring alternative
  • Easy to read and unbreakable
  • 1 year warranty
More About this Item
These self-adhesive reusable temperature labels provide a safe, accurate, easy-to-read means for monitoring temperatures. Indicating strips use liquid crystal technology to display the current temperature at an economical cost. The strip will turns a green color when ambient temperature is reached. Shelf life: 1 year after date of purchase.