Cole-Parmer Reversible 48-Point Vertical Magnetic Temperature Label, 26-120F; 10/Pk
Representative image only.
Cole-Parmer Item # WW-08078-48
Each style covers a wide range of temperatures, for use in many different application
  • Non-toxic / non-hazardous temperature measuring alternative
  • Easy to read and unbreakable
  • 1 year warranty
PRODUCT OPTIONS
₹11,100.00INR / Pkg of 10
Often Bought with…
SPECS & DESCRIPTION
Specifications & Description
  • Number of Temperature Indicators48
  • Max temperature (° F)120
  • Max temperature (° C)49
  • Temperature Points (° F)26 to 120°F in 2° increments
  • Temperature Points (° C)(-3), (-2), (-1), 1, 2, 3, 4, 6, 7, 8, 9, 10, 11, 12, 13, 14, 16, 17, 18, 19, 20, 21, 22, 23, 24, 26, 27, 28, 29, 30, 31, 32, 33, 34, 35, 37, 38, 39, 40, 41, 42, 43, 44, 45, 47, 48, 49
  • Initial colorBlack
  • First color changeGreen (actual temperature); Brown (above actual); Blue (below actual)
  • Second color changeBlack
  • Height (in)5
  • Width (in)1 1/2
  • Depth (in)3/64
  • DescriptionReversible 48-Point Vertical Magnetic Temperature Label, 26-120F; 10/Pk
  • Warranty1 year
1 Year Warranty
Key Features
  • Non-toxic / non-hazardous temperature measuring alternative
  • Easy to read and unbreakable
  • 1 year warranty
More About this Item
These self-adhesive reusable temperature labels provide a safe, accurate, easy-to-read means for monitoring temperatures. Indicating strips use liquid crystal technology to display the current temperature at an economical cost. The strip will turns a green color when ambient temperature is reached. Shelf life: 1 year after date of purchase.