Cole-Parmer Reversible 16-Point Vertical Temperature Label, 68-83C/154-184F; 10/Pk
Representative image only.
Cole-Parmer Item # WW-09037-23
Each style covers a wide range of temperatures, for use in many different application
  • Non-toxic / non-hazardous temperature measuring alternative
  • Easy to read and unbreakable
  • 1 year warranty
PRODUCT OPTIONS
₹2,100.00INR / Pkg of 10
SPECS & DESCRIPTION
Specifications & Description
  • Initial colorBlack
  • First color changeGreen (actual temperature); Brown (above actual); Blue (below actual)
  • DescriptionReversible 16-Point Vertical Temperature Label, 68-83C/154-184F; 10/Pk
  • Warranty1 year
1 Year Warranty
Key Features
  • Non-toxic / non-hazardous temperature measuring alternative
  • Easy to read and unbreakable
  • 1 year warranty
More About this Item
These self-adhesive reusable temperature labels provide a safe, accurate, easy-to-read means for monitoring temperatures. Indicating strips use liquid crystal technology to display the current temperature at an economical cost. The strip will turns a green color when ambient temperature is reached. Shelf life: 1 year after date of purchase.