Cole-Parmer Reusable Low-Temperature Labels, Refrigerator, 28 to 54F (-2 to 12C); 10/Pk
Representative image only.
Cole-Parmer Item # WW-90308-80
Ideal for use in refrigerator, freezer and cooler applications
  • Nontoxic nonhazardous temperature-measuring alternative
  • 1 year warranty
PRODUCT OPTIONS
₹6,000.00INR / Pkg of 10
SPECS & DESCRIPTION
Specifications & Description
  • Number of Temperature Indicators1
  • Max temperature (° F)54
  • Max temperature (° C)12
  • Temperature Points (° F)28 to 54
  • Temperature Points (° C)(-2 ) to (-12)
  • Height (in)5 1/2
  • Width (in)1
  • DescriptionReusable Low-Temperature Labels, Refrigerator, 28 to 54F (-2 to 12C); 10/Pk
  • Warranty1 year
1 Year Warranty
Key Features
  • Nontoxic nonhazardous temperature-measuring alternative
  • 1 year warranty
More About this Item
These self-adhesive reusable temperature labels provide a safe, accurate, easy-to-read means for monitoring low temperatures. Indicating strips use liquid crystal technology to display the current temperature range at an economical cost. Shelf life: 1 year after date of purchase.